Learn More
3-Diethylaminopropylamine, 99+%
CAS: 104-78-9 | C7H18N2 | 130.24 g/mol
Supplier: Thermo Scientific Chemicals 114090010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3-Diethylaminopropylamine | |
| 99.0 | |
| -60°C | |
| 0.8200g/mL | |
| 53°C | |
| 99% min. (GC) | |
| C7H18N2 | |
| (C2H5)2N(CH2)3NH2 | |
| 1 L | |
| 3-diethylaminopropylamine, 3-diethylamino propylamine, n,n-diethyl-1,3-propanediamine, 3-aminopropyl diethylamine, n1,n1-diethylpropane-1,3-diamine, 1,3-propanediamine, n,n-diethyl, diethylaminotrimethylenamine, n,n-diethyl-1,3-diaminopropane, 1-diethylamino propylamine-3, n-3-diethylaminopropyl amine | |
| QOHMWDJIBGVPIF-UHFFFAOYSA-N | |
| N',N'-diethylpropane-1,3-diamine | |
| 61011 | |
| 130.23 | |
| Liquid |
| 104-78-9 | |
| 100.0 | |
| Colorless to Yellow | |
| 168°C to 171°C | |
| Authentic | |
| Glass bottle | |
| 1.4406 to 1.4426 | |
| MFCD00008218 | |
| 0.82 | |
| Solubility in water: miscible with water | |
| CCN(CC)CCCN | |
| 130.24 | |
| 1.3 mPa.s (20°C) | |
| 99+% |
Chemical Identifiers
| 104-78-9 | |
| 130.24 | |
| QOHMWDJIBGVPIF-UHFFFAOYSA-N | |
| 61011 | |
| CCN(CC)CCCN |
| C7H18N2 | |
| MFCD00008218 | |
| 3-diethylaminopropylamine, 3-diethylamino propylamine, n,n-diethyl-1,3-propanediamine, 3-aminopropyl diethylamine, n1,n1-diethylpropane-1,3-diamine, 1,3-propanediamine, n,n-diethyl, diethylaminotrimethylenamine, n,n-diethyl-1,3-diaminopropane, 1-diethylamino propylamine-3, n-3-diethylaminopropyl amine | |
| N',N'-diethylpropane-1,3-diamine |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if swallowed.
Harmful in contact with skin.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-236-4
RUO – Research Use Only