missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[3-(Diethylamino)propyl]trimethoxysilane 97.0+%, TCI America™
$78.29 - $225.56
Chemical Identifiers
| CAS | 41051-80-3 |
|---|---|
| Molecular Formula | C10H25NO3Si |
| Molecular Weight (g/mol) | 235.40 |
| MDL Number | MFCD00039882 |
| InChI Key | ZLDHYRXZZNDOKU-UHFFFAOYSA-N |
| Synonym | N,N-Diethyl-3-(trimethoxysilyl)propylamine |
| PubChem CID | 170458 |
| IUPAC Name | diethyl[3-(trimethoxysilyl)propyl]amine |
| SMILES | CCN(CC)CCC[Si](OC)(OC)OC |
Chemical Identifiers
| 41051-80-3 | |
| 235.40 | |
| ZLDHYRXZZNDOKU-UHFFFAOYSA-N | |
| 170458 | |
| CCN(CC)CCC[Si](OC)(OC)OC |
| C10H25NO3Si | |
| MFCD00039882 | |
| N,N-Diethyl-3-(trimethoxysilyl)propylamine | |
| diethyl[3-(trimethoxysilyl)propyl]amine |
Specifications
| 41051-80-3 | |
| 120°C | |
| MFCD00039882 | |
| N,N-Diethyl-3-(trimethoxysilyl)propylamine | |
| CCN(CC)CCC[Si](OC)(OC)OC | |
| 235.40 | |
| 235.40 | |
| Liquid |
| Yellow | |
| C10H25NO3Si | |
| 5 mL | |
| ZLDHYRXZZNDOKU-UHFFFAOYSA-N | |
| diethyl[3-(trimethoxysilyl)propyl]amine | |
| 170458 | |
| ≥97.0% (GC,T) | |
| [3-(Diethylamino)propyl]trimethoxysilane |
Safety and Handling
TSCA : Yes