missing translation for 'onlineSavingsMsg'
Learn More

[3-(Diethylamino)propyl]trimethoxysilane 97.0+%, TCI America™

Catalog No. D43625ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43625ML 5 mL
D436225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43625ML Supplier TCI America Supplier No. D43625ML
Only null left

Chemical Identifiers

CAS 41051-80-3
Molecular Formula C10H25NO3Si
Molecular Weight (g/mol) 235.40
MDL Number MFCD00039882
InChI Key ZLDHYRXZZNDOKU-UHFFFAOYSA-N
Synonym N,N-Diethyl-3-(trimethoxysilyl)propylamine
PubChem CID 170458
IUPAC Name diethyl[3-(trimethoxysilyl)propyl]amine
SMILES CCN(CC)CCC[Si](OC)(OC)OC

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 mL
Formula Weight 235.40
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material [3-(Diethylamino)propyl]trimethoxysilane