missing translation for 'onlineSavingsMsg'
Learn More

3-Diethylamino-1-propyne, 98+%

CAS: 4079-68-9 | C7H13N | 111.188 g/mol

$92.68 - $370.71

Chemical Identifiers

CAS 4079-68-9
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.188
MDL Number MFCD00039881
InChI Key JZJXKEWVUBVOEH-UHFFFAOYSA-N
Synonym 1-diethylamino-2-propyne, diethylpropargylamine, 2-propyn-1-amine, n,n-diethyl, n,n-diethylpropargylamine, n,n-diethyl-2-propynylamine, 3-diethylamino propyne, 3-diethylamino-1-propyne, 3-n,n-diethylamino propyne, 1-n,n-diethylamino-2-propyne, 2-propynylamine, n,n-diethyl
PubChem CID 20010
IUPAC Name N,N-diethylprop-2-yn-1-amine
SMILES CCN(CC)CC#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1223706
View Documents
Thermo Scientific Chemicals
L1223706
5 g
Each for $92.68
Only null left
 
AAL1223714
View Documents
Thermo Scientific Chemicals
L1223714
25 g
Each for $370.71
Only null left
 
Description

Description

3-Diethylamino-1-propyne is used as functional leveling agent, brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Diethylamino-1-propyne is used as functional leveling agent, brightener.

Solubility
Soluble in acid. Insoluble in water.

Notes
Incompatible with heat, light, carbondioxide, heavy metal powders. Air & Light Sensitive. Store in the dark. Store under dry inert gas.
Specifications

Chemical Identifiers

4079-68-9
111.188
JZJXKEWVUBVOEH-UHFFFAOYSA-N
20010
CCN(CC)CC#C
C7H13N
MFCD00039881
1-diethylamino-2-propyne, diethylpropargylamine, 2-propyn-1-amine, n,n-diethyl, n,n-diethylpropargylamine, n,n-diethyl-2-propynylamine, 3-diethylamino propyne, 3-diethylamino-1-propyne, 3-n,n-diethylamino propyne, 1-n,n-diethylamino-2-propyne, 2-propynylamine, n,n-diethyl
N,N-diethylprop-2-yn-1-amine

Specifications

4079-68-9
119°C to 120°C
Amine-like
1.4305
MFCD00039881
UN2733
Air and light sensitive
Soluble in acid. Insoluble in water.
CCN(CC)CC#C
111.188
111.19
3-Diethylamino-1-propyne
0.804
20°C (68°F)
C7H13N
(CH3CH2)2NCH2C≡CH
5 g
1098468
1-diethylamino-2-propyne, diethylpropargylamine, 2-propyn-1-amine, n,n-diethyl, n,n-diethylpropargylamine, n,n-diethyl-2-propynylamine, 3-diethylamino propyne, 3-diethylamino-1-propyne, 3-n,n-diethylamino propyne, 1-n,n-diethylamino-2-propyne, 2-propynylamine, n,n-diethyl
JZJXKEWVUBVOEH-UHFFFAOYSA-N
N,N-diethylprop-2-yn-1-amine
20010
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H311-H314-H318-H302-H332
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H311-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

EINECSNumber : 223-804-5

RTECSNumber : UK5775000

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only