missing translation for 'onlineSavingsMsg'
Learn More

3-Diethylamino-1-propyne, 98+%

Catalog No. AAL1223706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1223706 5 g
AAL1223714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1223706 Supplier Thermo Scientific Chemicals Supplier No. L1223706

CAS: 4079-68-9 | C7H13N | 111.188 g/mol

3-Diethylamino-1-propyne is used as functional leveling agent, brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Diethylamino-1-propyne is used as functional leveling agent, brightener.

Solubility
Soluble in acid. Insoluble in water.

Notes
Incompatible with heat, light, carbondioxide, heavy metal powders. Air & Light Sensitive. Store in the dark. Store under dry inert gas.

Chemical Identifiers

CAS 4079-68-9
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.188
MDL Number MFCD00039881
InChI Key JZJXKEWVUBVOEH-UHFFFAOYSA-N
Synonym 1-diethylamino-2-propyne, diethylpropargylamine, 2-propyn-1-amine, n,n-diethyl, n,n-diethylpropargylamine, n,n-diethyl-2-propynylamine, 3-diethylamino propyne, 3-diethylamino-1-propyne, 3-n,n-diethylamino propyne, 1-n,n-diethylamino-2-propyne, 2-propynylamine, n,n-diethyl
PubChem CID 20010
IUPAC Name N,N-diethylprop-2-yn-1-amine
SMILES CCN(CC)CC#C

Specifications

Density 0.804
Boiling Point 119°C to 120°C
Flash Point 20°C (68°F)
Odor Amine-like
Refractive Index 1.4305
Linear Formula (CH3CH2)2NCH2C≡CH
Quantity 5 g
UN Number UN2733
Beilstein 1098468
Sensitivity Air and light sensitive
Solubility Information Soluble in acid. Insoluble in water.
Formula Weight 111.19
Percent Purity ≥98%
Chemical Name or Material 3-Diethylamino-1-propyne
Show More Show Less
Hazard Category H225-H302-H311-H314-H335
Hazard Statement GHS H Statement
H225-H311-H314-H318-H302-H332
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 223-804-5
RTECSNumber UK5775000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only