missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Diethylamino)-1,2-propanediol 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 233°C |
| Quantity | 500 g |
| UN Number | 2735 |
| Formula Weight | 147.22 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-(Diethylamino)-1,2-propanediol |
Safety and Handling
| EINECSNumber | (2)-3371 |
| TSCA | Yes |