missing translation for 'onlineSavingsMsg'
Learn More

3-(Diethylamino)-1,2-propanediol 98.0+%, TCI America™

Catalog No. D172125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D172125G 25 g
D1721500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D172125G Supplier TCI America Supplier No. D172125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 621-56-7
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.218
MDL Number MFCD00004716
InChI Key LTACQVCHVAUOKN-UHFFFAOYSA-N
Synonym 3-diethylamino-1,2-propanediol, 3-diethylamino propane-1,2-diol, 1,2-propanediol, 3-diethylamino, 3-diethylamino propan-1,2-diol, ksc496i7r, ltacqvchvauokn-uhfffaoysa, n,n-diethyl-3-amino-1,2-propanediol
PubChem CID 79074
IUPAC Name 3-(diethylamino)propane-1,2-diol
SMILES CCN(CC)CC(CO)O

Specifications

Color Yellow
Boiling Point 233°C
Quantity 25 g
UN Number 2735
Formula Weight 147.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Diethylamino)-1,2-propanediol
EINECSNumber (2)-3371
TSCA Yes