missing translation for 'onlineSavingsMsg'
Learn More

3-Diethylamino-1,2-propanediol, 97%

Catalog No. AAL0794322
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0794322 100 g
AAL0794314 25 g
AAL0794336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0794322 Supplier Thermo Scientific Chemicals Supplier No. L0794322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 621-56-7 | C7H17NO2 | 147.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-56-7
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.218
MDL Number MFCD00004716
InChI Key LTACQVCHVAUOKN-UHFFFAOYSA-N
Synonym 3-diethylamino-1,2-propanediol, 3-diethylamino propane-1,2-diol, 1,2-propanediol, 3-diethylamino, 3-diethylamino propan-1,2-diol, ksc496i7r, ltacqvchvauokn-uhfffaoysa, n,n-diethyl-3-amino-1,2-propanediol
PubChem CID 79074
IUPAC Name 3-(diethylamino)propane-1,2-diol
SMILES CCN(CC)CC(CO)O

Specifications

Density 0.97
Boiling Point 233°C to 235°C
Flash Point 107°C (224°F)
Refractive Index 1.4602
Quantity 100 g
UN Number UN2735
Beilstein 1737402
Formula Weight 147.22
Percent Purity 97%
Chemical Name or Material 3-Diethylamino-1,2-propanediol
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 210-693-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only