missing translation for 'onlineSavingsMsg'
Learn More

3-Diethoxyphosphoryloxy-1,2,3-benzotriazin-4(3H)-one, 98%

Catalog No. p-7029049
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2648106 5 g
AAH2648114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2648106 Supplier Thermo Scientific Chemicals Supplier No. H2648106
Only null left

CAS: 165534-43-0 | C11H14N3O5P | 299.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 165534-43-0
Molecular Formula C11H14N3O5P
Molecular Weight (g/mol) 299.223
MDL Number MFCD01236967
InChI Key AJDPNPAGZMZOMN-UHFFFAOYSA-N
Synonym depbt, 3-diethoxyphosphoryloxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxyphosphoryloxy-1,2,3-benzotrizin-4 3h-one, diethyl 4-oxobenzo d 1,2,3 triazin-3 4h-yl phosphate, diethyl 4-oxo-1,2,3-benzotriazin-3-yl phosphate, 3-diethoxyphosphoryloxy-3h-benzo d 1,2,3 triazin-4-one, 3-diethoxyphosphinyl oxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxyphosphoryl oxy-1,2,3-benzotriazin-4 3h-one, 3-diethoxy-phosphoryloxy-3h-benzo d 1,2,3 triazin-4-one
PubChem CID 4293995
IUPAC Name diethyl (4-oxo-1,2,3-benzotriazin-3-yl) phosphate
SMILES CCOP(=O)(OCC)ON1C(=O)C2=CC=CC=C2N=N1

Specifications

Melting Point 71°C to 76°C
Quantity 5 g
Formula Weight 299.22
Percent Purity 98%
Chemical Name or Material 3-Diethoxyphosphoryloxy-1,2,3-benzotriazin-4(3H)-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only