missing translation for 'onlineSavingsMsg'
Learn More

3-(Dicyanomethylidene)indan-1-one 98.0+%, TCI America™

Catalog No. p-7127087
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20541G 1 g
D20545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20541G Supplier TCI America Supplier No. D20541G
Only null left

Chemical Identifiers

CAS 1080-74-6
Molecular Formula C12H6N2O
Molecular Weight (g/mol) 194.19
MDL Number MFCD00143117
InChI Key QNVKZKOSAXYVFZ-UHFFFAOYSA-N
PubChem CID 3293617
IUPAC Name 2-(3-oxo-2,3-dihydro-1H-inden-1-ylidene)propanedinitrile
SMILES O=C1CC(=C(C#N)C#N)C2=CC=CC=C12

Specifications

Melting Point 228°C
Quantity 1 g
UN Number 2811
Formula Weight 194.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Dicyanomethylidene)indan-1-one