missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Dicyanomethylidene)indan-1-one 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 228°C |
| Quantity | 5 g |
| UN Number | 2811 |
| Formula Weight | 194.19 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-(Dicyanomethylidene)indan-1-one |
Safety and Handling
| TSCA | No |