missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3'-Deoxyuridine

Catalog No. AAJ65531MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65531MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65531MB Supplier Thermo Scientific Chemicals Supplier No. J65531MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7057-27-4 | C9H12N2O5 | 228.204 g/mol

A potential anti-cancer and anti-viral agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7057-27-4
Molecular Formula C9H12N2O5
Molecular Weight (g/mol) 228.204
MDL Number MFCD00079153
InChI Key QOXJRLADYHZRGC-SHYZEUOFSA-N
Synonym 3'-deoxyuridine, uridine, 3'-deoxy, 1-2r,3r,5s-3-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, 1-2r,3r,5s-3-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 3-deoxyuridine, 1-2r,3r,5s-3-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, uridine, 3/'-deoxy, 1-2r,3r,5s-3-hydroxy-5-hydroxymethyl oxolan-2-yl pyrimidine-2,4-dione
PubChem CID 145984
IUPAC Name 1-[(2R,3R,5S)-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1C(OC(C1O)N2C=CC(=O)NC2=O)CO

Specifications

Color White
Quantity 25 mg
Solubility Information Soluble in DMSO,methanol,or water; Also soluble in dichloromethane /n
Formula Weight 228.21
Physical Form Powder
Chemical Name or Material 3'-Deoxyuridine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only