missing translation for 'onlineSavingsMsg'
Learn More

3-Decyn-1-ol 97.0+%, TCI America™

Catalog No. D12755ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12755ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12755ML Supplier TCI America Supplier No. D12755ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51721-39-2
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00040917
InChI Key YGEQBZUDPQQIFI-UHFFFAOYSA-N
Synonym 3-decyn-1-ol, 2-hydroxyethylhexylacetylene, 3-decynol, pubchem13074, acmc-1b1ia, hexyl 2-hydroxyethyl acetylene, 2-hydroxyethyl-n-hexylacetylene
PubChem CID 103940
IUPAC Name dec-3-yn-1-ol
SMILES CCCCCCC#CCCO

Specifications

Color Yellow
Boiling Point 131°C
Quantity 5 mL
Formula Weight 154.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Decyn-1-ol