missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentylpropionyl chloride, 98%

Catalog No. AAA1771514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1771514 25 g
AAA1771522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1771514 Supplier Thermo Scientific Chemicals Supplier No. A1771514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 104-97-2 | C8H13ClO | 160.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-97-2
Molecular Formula C8H13ClO
Molecular Weight (g/mol) 160.64
MDL Number MFCD00001391
InChI Key SZQVEGOXJYTLLB-UHFFFAOYSA-N
Synonym 3-cyclopentylpropionyl chloride, cyclopentylpropionyl chloride, cyclopentanepropionyl chloride, 3-cyclopentyl propionyl chloride, cyclopentanepropanoyl-chloride, pubchem21363, cyclopentylpropionylchloride, cyclopentanepropanoylchloride, 3-cyclopentylpropanyl chloride, cyclopentane propionyl chloride
PubChem CID 66039
IUPAC Name 3-cyclopentylpropanoyl chloride
SMILES ClC(=O)CCC1CCCC1

Specifications

Density 1.049
Boiling Point 199°C to 200°C
Flash Point 84°C (184°F)
Odor Acrid
Refractive Index 1.4645
Quantity 25 g
UN Number UN3265
Beilstein 1856952
Sensitivity Moisture sensitive
Formula Weight 160.64
Percent Purity 98%
Chemical Name or Material 3-Cyclopentylpropionyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 203-257-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only