missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentylpropionic acid, 98%

CAS: 140-77-2 | C8H14O2 | 142.20 g/mol

Supplier:  Thermo Scientific Chemicals A1819222

 View more versions of this product

Catalog No. AAA1819222


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Cyclopentylpropionic acid
0.996
47°C (117°F)
1.457
100 g
3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate
OC(=O)CCC1CCCC1
142.20
CHEBI:50899
98%
140-77-2
130°C to 133°C (12 mmHg)
C8H14O2
MFCD00001392
UN1993
ZRPLANDPDWYOMZ-UHFFFAOYSA-N
3-cyclopentylpropanoic acid
8818
142.2

Chemical Identifiers

140-77-2
142.20
ZRPLANDPDWYOMZ-UHFFFAOYSA-N
8818
3-cyclopentylpropanoic acid
C8H14O2
MFCD00001392
3-cyclopentylpropionic acid, cyclopentanepropanoic acid, cyclopentanepropionic acid, 3-cylcopentylpropanoic acid, cyclopentylpropionic acid, propionic acid, 3-cyclopentyl, 3-cyclopentyl-propionic acid, cypionic acid, 3-cyclopentylpropionicacid, cipionate
CHEBI:50899
OC(=O)CCC1CCCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 205-433-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only