missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexylthiophene 96.0+%, TCI America™

Catalog No. C34001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34001G 1 g
C34005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34001G Supplier TCI America Supplier No. C34001G
Only null left

Chemical Identifiers

CAS 120659-34-9
Molecular Formula C10H14S
Molecular Weight (g/mol) 166.282
MDL Number MFCD02260242
InChI Key FGHYZEFIPLFAOC-UHFFFAOYSA-N
PubChem CID 1519411
IUPAC Name 3-cyclohexylthiophene
SMILES C1CCC(CC1)C2=CSC=C2

Specifications

Quantity 1 g
Formula Weight 166.28
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Cyclohexylthiophene
TSCA No
Recommended Storage Refrigerator