missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexylaminopropanesulfonic Acid 98.0+%, TCI America™

Catalog No. p-7124684
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C092125G 25 g
C0921250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C092125G Supplier TCI America Supplier No. C092125G
Only null left

Chemical Identifiers

CAS 1135-40-6
Molecular Formula C9H19NO3S
Molecular Weight (g/mol) 221.32
MDL Number MFCD00003837
InChI Key PJWWRFATQTVXHA-UHFFFAOYSA-N
Synonym caps, 3-cyclohexylaminopropanesulfonic acid, n-cyclohexyl-3-aminopropanesulfonic acid, 3-cyclohexylamino-1-propanesulfonic acid, 1-propanesulfonic acid, 3-cyclohexylamino, 3-cyclohexylamino propane-1-sulfonic acid, unii-4w981o1lxp, 3-cyclohexylamino propanesulfonic acid, 3-cyclohexylaminopropane-1-sulphonic acid, 3-cyclohexyl-1-propylsulfonic acid
PubChem CID 70815
IUPAC Name 3-(cyclohexylamino)propane-1-sulfonic acid
SMILES OS(=O)(=O)CCCNC1CCCCC1

Specifications

Color White
Quantity 25 g
Formula Weight 221.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyclohexylaminopropanesulfonic Acid [Good′s buffer component for biological research]
RTECSNumber TZ6395000
TSCA Yes