missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexyl-1-propyne, 97%

Catalog No. AAB2050503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2050503 1 g
AAB2050506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2050503 Supplier Thermo Scientific Chemicals Supplier No. B2050503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17715-00-3 | C9H14 | 122.211 g/mol

3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyclohexyl-1-propyne, is used as a pharmaceutical intermediate. It can be used to produce (5-bromo-4,4,5,5-tetrafluoro-pent-2-enyl)-cyclohexane at the temperature of 40°C. It will need reagents (NH4)2S2O8, HCO2Na·2H2O and solvent dimethylformamide with the reaction time of 3 hours. It is also used as organic synthesis intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 17715-00-3
Molecular Formula C9H14
Molecular Weight (g/mol) 122.211
MDL Number MFCD00041562
InChI Key UARFKZSJGDQRLF-UHFFFAOYSA-N
Synonym 3-cyclohexyl-1-propyne, prop-2-yn-1-ylcyclohexane, 1-prop-2-ynylcyclohexane, 3-cyclohexylpropyne, 2-propynylcyclohexane, propyne, 3-cyclohexyl, cyclohexane, 2-propynyl-9ci, prop-2-ynyl-cyclohexane, cyclohexane, 2-propynyl, 3-cyclohexylprop-1-yne
PubChem CID 87265
IUPAC Name prop-2-ynylcyclohexane
SMILES C#CCC1CCCCC1

Specifications

Density 0.845
Boiling Point 157°C to 158°C
Flash Point 35°C (95°F)
Refractive Index 1.462
Quantity 1 g
UN Number UN3295
Beilstein 1849402
Solubility Information Slightly soluble in water.
Formula Weight 122.21
Percent Purity 97%
Chemical Name or Material 3-Cyclohexyl-1-propyne
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
RTECSNumber UK4725000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only