missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyclohexene-1-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America C0652100G
Specifications
| 3-Cyclohexene-1-carboxylic Acid | |
| Yellow | |
| C7H10O2 | |
| 100 g | |
| 3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid | |
| C1CC(CC=C1)C(=O)O | |
| 126.155 | |
| 126.16 | |
| Liquid |
| 4771-80-6 | |
| 238°C | |
| MFCD00013781 | |
| 3265 | |
| VUSWCWPCANWBFG-UHFFFAOYSA-N | |
| cyclohex-3-ene-1-carboxylic acid | |
| 20903 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 4771-80-6 | |
| 126.155 | |
| VUSWCWPCANWBFG-UHFFFAOYSA-N | |
| 20903 | |
| C1CC(CC=C1)C(=O)O |
| C7H10O2 | |
| MFCD00013781 | |
| 3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid | |
| cyclohex-3-ene-1-carboxylic acid |
Safety and Handling
RTECSNumber : GW3675000
TSCA : Yes