missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexene-1-carboxylic Acid 98.0+%, TCI America™

Catalog No. C0652500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0652500G 500 g
C065225G 25 g
C0652100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0652500G Supplier TCI America Supplier No. C0652500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4771-80-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00013781
InChI Key VUSWCWPCANWBFG-UHFFFAOYSA-N
Synonym 3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid
PubChem CID 20903
IUPAC Name cyclohex-3-ene-1-carboxylic acid
SMILES C1CC(CC=C1)C(=O)O

Specifications

Color Yellow
Boiling Point 238°C
Quantity 500 g
UN Number 3265
Formula Weight 126.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Cyclohexene-1-carboxylic Acid
RTECSNumber GW3675000
TSCA Yes