missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexene-1-carboxylic acid, 98%

Catalog No. AAA1522914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1522914 25 g
AAA1522906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1522914 Supplier Thermo Scientific Chemicals Supplier No. A1522914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4771-80-6 | C7H10O2 | 126.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4771-80-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00013781
InChI Key VUSWCWPCANWBFG-UHFFFAOYSA-N
Synonym 3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid
PubChem CID 20903
IUPAC Name cyclohex-3-ene-1-carboxylic acid
SMILES C1CC(CC=C1)C(=O)O

Specifications

Melting Point 16°C to 18°C
Density 1.082
Boiling Point 120°C to 122°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4815
Quantity 25 g
UN Number UN3265
Beilstein 1617723
Solubility Information V sol
Formula Weight 126.16
Percent Purity 98%
Chemical Name or Material 3-Cyclohexene-1-carboxylic acid
Show More Show Less
Hazard Category H312-H314-H335
Hazard Statement GHS H Statement
H311-H314-H318
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 225-314-7
RTECSNumber GW3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only