missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexene-1-carboxaldehyde 98.0+%, TCI America™

Supplier:  TCI America C088125ML

Catalog No. C088125ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Cyclohexene-1-carboxaldehyde
Colorless
C7H10O
25 mL
3-cyclohexene-1-carboxaldehyde, 1,2,3,6-tetrahydrobenzaldehyde, 4-formylcyclohexene, cyclohexene-4-carboxaldehyde, 1-formyl-3-cyclohexene, 3-cyclohexen-1-aldehyde, 1-cyclohexene-4-carboxaldehyde, 1,2,5,6-tetrahydrobenzaldehyde, cyclohex-3-enecarbaldehyde, tetrahydrobenzaldehyde
O=CC1CCC=CC1
110.16
110.16
Liquid
100-50-5
164°C
MFCD00001572
2498
DCFDVJPDXYGCOK-UHFFFAOYNA-N
cyclohex-3-ene-1-carbaldehyde
7508
≥98.0% (GC)

Chemical Identifiers

100-50-5
110.16
DCFDVJPDXYGCOK-UHFFFAOYNA-N
7508
O=CC1CCC=CC1
C7H10O
MFCD00001572
3-cyclohexene-1-carboxaldehyde, 1,2,3,6-tetrahydrobenzaldehyde, 4-formylcyclohexene, cyclohexene-4-carboxaldehyde, 1-formyl-3-cyclohexene, 3-cyclohexen-1-aldehyde, 1-cyclohexene-4-carboxaldehyde, 1,2,5,6-tetrahydrobenzaldehyde, cyclohex-3-enecarbaldehyde, tetrahydrobenzaldehyde
cyclohex-3-ene-1-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3656

RTECSNumber : GW2800000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions