missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanoumbelliferone 98.0+%, TCI America™

Catalog No. C28341G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28341G 1 g
C28345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28341G Supplier TCI America Supplier No. C28341G
Only null left

Chemical Identifiers

CAS 19088-73-4
Molecular Formula C10H5NO3
Molecular Weight (g/mol) 187.15
MDL Number MFCD00037480
InChI Key IJQYTHQDUDCJEQ-UHFFFAOYSA-N
Synonym 3-Cyano-7-hydroxycoumarin
PubChem CID 5393173
IUPAC Name 7-hydroxy-2-oxo-2H-chromene-3-carbonitrile
SMILES OC1=CC=C2C=C(C#N)C(=O)OC2=C1

Specifications

Melting Point 280°C
Color Yellow
Quantity 1 g
Formula Weight 187.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyanoumbelliferone