missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanothiophene 98.0+%, TCI America™

Catalog No. C13445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13445G 5 g
C134425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13445G Supplier TCI America Supplier No. C13445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1641-09-4
Molecular Formula C5H3NS
Molecular Weight (g/mol) 109.146
MDL Number MFCD00151852
InChI Key GSXCEVHRIVLFJV-UHFFFAOYSA-N
Synonym 3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h
PubChem CID 74231
IUPAC Name thiophene-3-carbonitrile
SMILES C1=CSC=C1C#N

Specifications

Color Yellow
Boiling Point 205°C
Quantity 5 g
Formula Weight 109.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Cyanothiophene