missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridine 1-oxide, 97%

Catalog No. AAA1996188
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1996188 2.5 g
AAA1996109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1996188 Supplier Thermo Scientific Chemicals Supplier No. A1996188
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14906-64-0 | C6H5N2O | 121.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14906-64-0
Molecular Formula C6H5N2O
Molecular Weight (g/mol) 121.12
MDL Number MFCD00129037
InChI Key AABSWFBNBQBPEQ-UHFFFAOYNA-N
Synonym nicotinonitrile-1-oxide, 3-cyanopyridine n-oxide, 3-cyanopyridine-n-oxide, nicotinonitrile 1-oxide, 3-cyanopyridine 1-oxide, 3-cyanopyridin-1-ium-1-olate, 3-pyridinecarbonitrile, 1-oxide, acmc-1c6hr, 3-pyridinecarbonitrile,1-oxide, woovsqcalyyudo-uhfffaoysa
PubChem CID 84686
SMILES [O-][NH++]1C=C[CH-]C(=C1)C#N

Specifications

Melting Point 175°C to 178°C
Quantity 2.5 g
Sensitivity Hygroscopic
IUPAC Name 1-oxidopyridin-1-ium-3-carbonitrile
Formula Weight 120.11
Percent Purity 97%
Chemical Name or Material 3-Cyanopyridine 1-oxide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 238-975-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only