missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridazine, 97%

Catalog No. p-7032194
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33568MD 250 mg
AAH3356803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33568MD Supplier Thermo Scientific Chemicals Supplier No. H33568MD
Only null left

CAS: 53896-49-4 | C5H3N3 | 105.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53896-49-4
Molecular Formula C5H3N3
Molecular Weight (g/mol) 105.1
MDL Number MFCD09881239
InChI Key PJESVVYWPFAJCS-UHFFFAOYSA-N
Synonym 3-cyanopyridazine, 3-pyridazinecarbonitrile, 3-cyano-1,2-diazine, pyridazine-3-carbonitrile
PubChem CID 13642940
IUPAC Name pyridazine-3-carbonitrile
SMILES C1=CC(=NN=C1)C#N

Specifications

Melting Point 43°C to 45°C
Quantity 250 mg
UN Number UN3439
Formula Weight 105.1
Percent Purity 97%
Chemical Name or Material 3-Cyanopyridazine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only