missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopropylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58962AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58962AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58962AD Supplier Thermo Scientific Chemicals Supplier No. H58962AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 135579-85-0 | C4H6BrNZn | 213.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-vent

Chemical Identifiers

CAS 135579-85-0
Molecular Formula C4H6BrNZn
Molecular Weight (g/mol) 213.38
MDL Number MFCD00671975
InChI Key HEHCQTGEYNYOSF-UHFFFAOYSA-M
Synonym 3-cyanopropylzinc bromide solution, 3-cyanopropylzinc bromide, 4-bromozincio butanenitrile, 3-cyanopropylzinc bromide 0.5 m in tetrahydrofuran, 3-cyanopropylzinc bromide solution, 0.5 m in thf, 3-cyanopropylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 4129356
IUPAC Name bromozinc(1+);butanenitrile
SMILES [Zn+]Br.[CH2-]CCC#N

Specifications

Density 0.981
Linear Formula N≡C(CH2)3ZnBr
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Solubility Information Reacts with water.
Formula Weight 213.38
Chemical Name or Material 3-Cyanopropylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H300-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Fatal if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only