missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopropyldimethylchlorosilane, 95%

Catalog No. AC313380100
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313380100 10 mL Glass bottle
AC313380500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC313380100 Supplier Thermo Scientific Chemicals Supplier No. 313380100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18156-15-5 | C6H12ClNSi | 161.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18156-15-5
Molecular Formula C6H12ClNSi
Molecular Weight (g/mol) 161.7
InChI Key GPIARXZSVWTOMD-UHFFFAOYSA-N
Synonym 3-cyanopropyl dimethylchlorosilane, 4-chlorodimethylsilyl butanenitrile, 3-cyanopropyldimethylchlorosilane, chloro 3-cyanopropyl dimethylsilane, butanenitrile, 4-chlorodimethylsilyl, 4-chlorodimethylsilyl butyronitrile, 4-chloro dimethyl silyl butanenitrile, cyanopropyldimethylchlorosilane, chlorocyanopropyldimethylsilane, cyanopropyl dimethylchlorosilane
PubChem CID 87478
IUPAC Name 4-[chloro(dimethyl)silyl]butanenitrile
SMILES C[Si](C)(CCCC#N)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown to Colorless
Density 0.9900g/mL
Boiling Point 230.0°C to 235.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4450 to 1.4470 (20°C, 589nm)
Linear Formula (CH3)2Si(Cl)(CH2)3CN
Quantity 10 mL
Specific Gravity 0.99
Solubility Information Solubility in water: reacts.
Formula Weight 161.7
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Cyanopropyldimethylchlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 242-039-8

RUO – Research Use Only