missing translation for 'onlineSavingsMsg'
Learn More

(3-Cyanopropyl)trichlorosilane, 98%

Catalog No. p-7021452
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1378714 25 g
AAA1378722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1378714 Supplier Thermo Scientific Chemicals Supplier No. A1378714

CAS: 1071-27-8 | C4H6Cl3NSi | 202.534 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1071-27-8
Molecular Formula C4H6Cl3NSi
Molecular Weight (g/mol) 202.534
MDL Number MFCD00013832
InChI Key HMFFOEBLYHLRQN-UHFFFAOYSA-N
Synonym 3-cyanopropyltrichlorosilane, butanenitrile, 4-trichlorosilyl, 4-trichlorosilyl butyronitrile, 4-cyanobutyltrichlorosilane, butyronitrile, 4-trichlorosilyl, trichloro 3-cyanopropyl silane, trichlor-3-kyanpropylsilan, silane, trichloro 3-cyanopropyl, unii-05m8iu15r8, gamma-cyanopropyltrichlorosilane
PubChem CID 14063
IUPAC Name 4-trichlorosilylbutanenitrile
SMILES C(CC#N)C[Si](Cl)(Cl)Cl

Specifications

Boiling Point 237°C to 238°C
Quantity 25 g
Linear Formula Cl3Si(CH2)3CN
UN Number UN2987
Beilstein 906924
Flash Point 92°C (198°F)
Formula Weight 202.54
Percent Purity 98%
Odor Pungent
Refractive Index 1.463
Sensitivity Moisture sensitive
Density 1.28
Chemical Name or Material (3-Cyanopropyl)trichlorosilane
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber 213-990-6
RTECSNumber VV2775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only