missing translation for 'onlineSavingsMsg'
Learn More

(3-Cyanopropyl)dimethylchlorosilane 95.0+%, TCI America™

Catalog No. c120725ml
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C120725ML 25 mL
C12075ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C120725ML Supplier TCI America Supplier No. C120725ML

Chemical Identifiers

CAS 18156-15-5
Molecular Formula C6H12ClNSi
Molecular Weight (g/mol) 161.704
MDL Number MFCD00001969
InChI Key GPIARXZSVWTOMD-UHFFFAOYSA-N
Synonym 3-cyanopropyl dimethylchlorosilane, 4-chlorodimethylsilyl butanenitrile, 3-cyanopropyldimethylchlorosilane, chloro 3-cyanopropyl dimethylsilane, butanenitrile, 4-chlorodimethylsilyl, 4-chlorodimethylsilyl butyronitrile, 4-chloro dimethyl silyl butanenitrile, cyanopropyldimethylchlorosilane, chlorocyanopropyldimethylsilane, cyanopropyl dimethylchlorosilane
PubChem CID 87478
IUPAC Name 4-[chloro(dimethyl)silyl]butanenitrile
SMILES C[Si](C)(CCCC#N)Cl

Specifications

Color Yellow
Boiling Point 109°C
Quantity 25 mL
UN Number 2987
Formula Weight 161.70
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (3-Cyanopropyl)dimethylchlorosilane