missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanophenylboronic acid pinacol ester, 97%

Catalog No. p-4228556
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433790010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433790010 Supplier Thermo Scientific Chemicals Supplier No. 433790010
Only null left

CAS: 214360-46-0 | C13H16BNO2 | 229.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 214360-46-0
Molecular Formula C13H16BNO2
Molecular Weight (g/mol) 229.09
MDL Number MFCD03789262
InChI Key FIGQEPXOSAFKTA-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyanophenylboronic acid pinacol ester, 3-cyanophenylboronic acid, pinacol ester, benzonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-cyanobenzeneboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, pubchem15603, 3-cyanophenylboronicacidpinacolester, 3-cyanobenzeneboronic acid, pinacol ester
PubChem CID 2760590
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 80.0°C to 81.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 229.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyanophenylboronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only