missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyanophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$93.21 - $93.21
Chemical Identifiers
| CAS | 150255-96-2 |
|---|---|
| Molecular Formula | C7H6BNO2 |
| Molecular Weight (g/mol) | 146.94 |
| MDL Number | MFCD01318967 |
| InChI Key | XDBHWPLGGBLUHH-UHFFFAOYSA-N |
| PubChem CID | 2734325 |
| IUPAC Name | (3-cyanophenyl)boronic acid |
| SMILES | OB(O)C1=CC=CC(=C1)C#N |
Chemical Identifiers
| 150255-96-2 | |
| 146.94 | |
| XDBHWPLGGBLUHH-UHFFFAOYSA-N | |
| (3-cyanophenyl)boronic acid |
| C7H6BNO2 | |
| MFCD01318967 | |
| 2734325 | |
| OB(O)C1=CC=CC(=C1)C#N |
Specifications
| 150255-96-2 | |
| White-Yellow | |
| MFCD01318967 | |
| 3439 | |
| XDBHWPLGGBLUHH-UHFFFAOYSA-N | |
| (3-cyanophenyl)boronic acid | |
| 2734325 | |
| Crystalline Powder |
| 298°C | |
| C7H6BNO2 | |
| 5 g | |
| 3-cyanophenyl boronic acid, 3-cyanobenzeneboronic acid, 3-boronobenzonitrile, 3-dihydroxyboranyl benzonitrile, 3-cyanobenzene boronic acid, 3-cyano-phenyl-boronic acid, boronic acid, 3-cyanophenyl, pubchem1804, phenylboronic acid, 4 | |
| OB(O)C1=CC=CC(=C1)C#N | |
| 146.94 | |
| 146.94 | |
| 3-Cyanophenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No