missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C20315G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20315G Supplier TCI America Supplier No. C20315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150255-96-2
Molecular Formula C7H6BNO2
Molecular Weight (g/mol) 146.94
MDL Number MFCD01318967
InChI Key XDBHWPLGGBLUHH-UHFFFAOYSA-N
PubChem CID 2734325
IUPAC Name (3-cyanophenyl)boronic acid
SMILES OB(O)C1=CC=CC(=C1)C#N

Specifications

Melting Point 298°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Synonym 3-cyanophenyl boronic acid, 3-cyanobenzeneboronic acid, 3-boronobenzonitrile, 3-dihydroxyboranyl benzonitrile, 3-cyanobenzene boronic acid, 3-cyano-phenyl-boronic acid, boronic acid, 3-cyanophenyl, pubchem1804, phenylboronic acid, 4
Formula Weight 146.94
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyanophenylboronic Acid (contains varying amounts of Anhydride)