missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanophenyl isocyanate, 97%

Catalog No. aal0943206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0943206 5 g
AAL0943203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0943206 Supplier Thermo Scientific Chemicals Supplier No. L0943206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16413-26-6 | C8H4N2O | 144.133 g/mol

3-Cyanophenyl isocyanate serves as a starting reagent in the synthesis of R/S-4-(3-chlorophenylaminocarbonylamino)-3,4-dihydro-2,2-dimethyl-2H-1-benzopyran and 1-(3-cyanophenyl)-3-(2-methoxy-6-pentadecylbenzyl)urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyanophenyl isocyanate serves as a starting reagent in the synthesis of R/S-4-(3-chlorophenylaminocarbonylamino)-3,4-dihydro-2,2-dimethyl-2H-1-benzopyran and 1-(3-cyanophenyl)-3-(2-methoxy-6-pentadecylbenzyl)urea.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 16413-26-6
Molecular Formula C8H4N2O
Molecular Weight (g/mol) 144.133
MDL Number MFCD00013856
InChI Key NZHPVPMRNASEQK-UHFFFAOYSA-N
Synonym 3-cyanophenyl isocyanate, 3-cyanophenylisocyanate, benzonitrile, 3-isocyanato, 3-isocyanatobenzenecarbonitrile, m-cyanophenyl isocyanate, m-cyanophenylisocyanate, pubchem5028, 3-isocyanato-benzonitrile, acmc-1c7a7, paragos 331038
PubChem CID 2733315
IUPAC Name 3-isocyanatobenzonitrile
SMILES C1=CC(=CC(=C1)N=C=O)C#N

Specifications

Melting Point 51°C to 54°C
Odor Irritating
Quantity 5 g
UN Number UN2811
Beilstein 3089117
Sensitivity Moisture sensitive
Formula Weight 144.13
Percent Purity 97%
Chemical Name or Material 3-Cyanophenyl isocyanate
Hazard Category H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H300-H310-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only