missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanophenyl isocyanate, 97%

CAS: 16413-26-6 | C8H4N2O | 144.13 g/mol

$190.32 - $190.32

Chemical Identifiers

CAS 16413-26-6
Molecular Formula C8H4N2O
Molecular Weight (g/mol) 144.13
MDL Number MFCD00013856
InChI Key NZHPVPMRNASEQK-UHFFFAOYSA-N
Synonym 3-cyanophenyl isocyanate, 3-cyanophenylisocyanate, benzonitrile, 3-isocyanato, 3-isocyanatobenzenecarbonitrile, m-cyanophenyl isocyanate, m-cyanophenylisocyanate, pubchem5028, 3-isocyanato-benzonitrile, acmc-1c7a7, paragos 331038
PubChem CID 2733315
IUPAC Name 3-isocyanatobenzonitrile
SMILES C1=CC(=CC(=C1)N=C=O)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC314180050
SDS View Documents
Thermo Scientific Chemicals
314180050
5 g Glass bottle
Each for $190.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16413-26-6
144.13
NZHPVPMRNASEQK-UHFFFAOYSA-N
2733315
C1=CC(=CC(=C1)N=C=O)C#N
C8H4N2O
MFCD00013856
3-cyanophenyl isocyanate, 3-cyanophenylisocyanate, benzonitrile, 3-isocyanato, 3-isocyanatobenzenecarbonitrile, m-cyanophenyl isocyanate, m-cyanophenylisocyanate, pubchem5028, 3-isocyanato-benzonitrile, acmc-1c7a7, paragos 331038
3-isocyanatobenzonitrile

Specifications

16413-26-6
100.0
White to Yellow
96% min. (GC)
C8H4N2O
MFCD00013856
3-cyanophenyl isocyanate, 3-cyanophenylisocyanate, benzonitrile, 3-isocyanato, 3-isocyanatobenzenecarbonitrile, m-cyanophenyl isocyanate, m-cyanophenylisocyanate, pubchem5028, 3-isocyanato-benzonitrile, acmc-1c7a7, paragos 331038
C1=CC(=CC(=C1)N=C=O)C#N
144.13
144.13
Low Melting Solid
96.0
49°C to 54°C
Authentic
Glass bottle
CNC6H4NCO
5 g
NZHPVPMRNASEQK-UHFFFAOYSA-N
3-isocyanatobenzonitrile
2733315
97%
3-Cyanophenyl isocyanate, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only