missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanophenacyl Bromide 98.0+%, TCI America™

Catalog No. B40795G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B40795G Supplier TCI America Supplier No. B40795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50916-55-7
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.06
MDL Number MFCD00833267
InChI Key XWCGNFLHRINYCE-UHFFFAOYSA-N
Synonym 3-2-bromoacetyl benzonitrile, 3-cyanophenacyl bromide, 3-cyanophenacylbromide, 2-bromo-3'-cyanoacetophenone, 3-2-bromoacetyl benzenecarbonitrile, 3-bromoacetyl benzonitrile, 3-2-bromo-acetyl-benzonitrile, benzonitrile, 3-bromoacetyl, 3-bromoacetyl-benzonitrile
PubChem CID 2735867
IUPAC Name 3-(2-bromoacetyl)benzonitrile
SMILES BrCC(=O)C1=CC=CC(=C1)C#N

Specifications

Melting Point 71°C
Quantity 5 g
UN Number 2923
Formula Weight 224.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyanophenacyl Bromide