missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Cyanomethyl)-2,4,5-trimethylthiophene 98.0+%, TCI America™
$102.52 - $503.30
Chemical Identifiers
| CAS | 112440-49-0 |
|---|---|
| Molecular Formula | C9H11NS |
| Molecular Weight (g/mol) | 165.25 |
| MDL Number | MFCD00143113 |
| InChI Key | GVRRCQCZKOPGDZ-UHFFFAOYSA-N |
| PubChem CID | 14122208 |
| IUPAC Name | 2-(2,4,5-trimethylthiophen-3-yl)acetonitrile |
| SMILES | CC1=C(C)C(CC#N)=C(C)S1 |
Chemical Identifiers
| 112440-49-0 | |
| 165.25 | |
| GVRRCQCZKOPGDZ-UHFFFAOYSA-N | |
| 2-(2,4,5-trimethylthiophen-3-yl)acetonitrile |
| C9H11NS | |
| MFCD00143113 | |
| 14122208 | |
| CC1=C(C)C(CC#N)=C(C)S1 |
Specifications
| 112440-49-0 | |
| White-Yellow | |
| C9H11NS | |
| 1 g | |
| CC1=C(C)C(CC#N)=C(C)S1 | |
| 165.25 | |
| 165.25 | |
| Crystalline Powder |
| 66°C | |
| 130°C | |
| MFCD00143113 | |
| GVRRCQCZKOPGDZ-UHFFFAOYSA-N | |
| 2-(2,4,5-trimethylthiophen-3-yl)acetonitrile | |
| 14122208 | |
| ≥98.0% (GC) | |
| 3-(Cyanomethyl)-2,4,5-trimethylthiophene |
Safety and Handling
TSCA : No