missing translation for 'onlineSavingsMsg'
Learn More

3-(Cyanomethyl)-2,4,5-trimethylthiophene 98.0+%, TCI America™

Catalog No. C12281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12281G 1 g
C12285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12281G Supplier TCI America Supplier No. C12281G

Chemical Identifiers

CAS 112440-49-0
Molecular Formula C9H11NS
Molecular Weight (g/mol) 165.25
MDL Number MFCD00143113
InChI Key GVRRCQCZKOPGDZ-UHFFFAOYSA-N
PubChem CID 14122208
IUPAC Name 2-(2,4,5-trimethylthiophen-3-yl)acetonitrile
SMILES CC1=C(C)C(CC#N)=C(C)S1

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 165.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Cyanomethyl)-2,4,5-trimethylthiophene