missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-5-methylpyridine, 98+%

Catalog No. p-7047037
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17427MD 250 mg
AAL1742703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL17427MD Supplier Thermo Scientific Chemicals Supplier No. L17427MD
Only null left

CAS: 42885-14-3 | C7H6N2 | 118.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42885-14-3
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00673151
InChI Key VDWQWOXVBXURMT-UHFFFAOYSA-N
Synonym 3-cyano-5-methylpyridine, 5-methylnicotinonitrile, unii-15462uxn1x, 5-methyl-nicotinonitrile, 5-methyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 5-methyl, 3-cyano-5-methylpyridine 3-cyano-5-picoline, 5-cyano-3-picoline, 5-cyano-3-methylpyridine, acmc-1ao5a
PubChem CID 637838
IUPAC Name 5-methylpyridine-3-carbonitrile
SMILES CC1=CN=CC(=C1)C#N

Specifications

Melting Point 82°C to 85°C
Quantity 250 mg
Beilstein 111258
Formula Weight 118.14
Percent Purity ≥98%
Chemical Name or Material 3-Cyano-5-methylpyridine
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 255-988-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only