missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-5-hydroxypyridine, 97%

Catalog No. AAH33006MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33006MD 250 mg
AAH3300603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33006MD Supplier Thermo Scientific Chemicals Supplier No. H33006MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 152803-24-2 | C6H4N2O | 120.11 g/mol

3-Cyano-5-hydroxypyridine is active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyano-5-hydroxypyridine is active pharmaceutical ingredients.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 152803-24-2
Molecular Formula C6H4N2O
Molecular Weight (g/mol) 120.11
MDL Number MFCD11044307
InChI Key KAXIYYPIORYZLB-UHFFFAOYSA-N
PubChem CID 15342142
IUPAC Name 5-hydroxypyridine-3-carbonitrile
SMILES OC1=CN=CC(=C1)C#N

Specifications

Melting Point 213°C to 217°C
Quantity 250 mg
UN Number UN3439
Synonym 5-hydroxynicotinonitrile, 3-cyano-5-hydroxypyridine, 3-pyridinecarbonitrile, 5-hydroxy, 5-hydroxy-nicotinonitrile, 5-hydroxy-3-pyridinecarbonitrile, 5-cyano-3-hydroxypyridine, pubchem17727, kaxiyypioryzlb-uhfffaoysa, 5-cyano-5-hydroxypyridine, 3-pyridinecarbonitrile,5-hydroxy
Solubility Information Slightly soluble in water.
Formula Weight 120.11
Percent Purity 97%
Chemical Name or Material 3-Cyano-5-hydroxypyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only