missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-methylpyridine 98.0+%, TCI America™

Catalog No. C26081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26081G 1 g
C26085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26081G Supplier TCI America Supplier No. C26081G
Only null left

Chemical Identifiers

CAS 5444-01-9
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00234272
InChI Key XLAPHZHNODDMDD-UHFFFAOYSA-N
Synonym 3-cyano-4-methylpyridine, 4-methylnicotinonitrile, 3-cyano-4-picoline, 4-methyl-nicotinonitrile, 3-pyridinecarbonitrile, 4-methyl, 4-methyl-3-pyridinecarbonitrile, 3-cyano-4-methylpyridine 3-cyano-4-picoline, pubchem6623, pubchem18426, 4-methyl nicotinonitrile
PubChem CID 227806
IUPAC Name 4-methylpyridine-3-carbonitrile
SMILES CC1=C(C=NC=C1)C#N

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 111°C
Quantity 1 g
Formula Weight 118.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyano-4-methylpyridine