missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyano-4-methoxy-2-pyridone 98.0+%, TCI America™
Supplier: TCI America C292525G
Specifications
| 3-Cyano-4-methoxy-2-pyridone | |
| 278°C | |
| C7H6N2O2 | |
| 25 g | |
| 4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile, 4-methoxy-2-oxo-1,2-dihydro-pyridine-3-carbonitrile, 3-cyano-2-hydroxy-4-methoxypyridine, 4-methoxy-2-oxo-1,2-dihydro-3-pyridinecarbonitrile, 2-hydroxy-4-methoxynicotinonitrile, 1,2-dihydro-4-methoxy-2-oxo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4-methoxy-2-oxo, 3-cyano-4-methoxy-2 1h-pyridinone, 2-hydroxy-4-methoxy-nicotinonitrile, 3-cyano-4-methoxy-2-1h-pyridinone | |
| COC1=C(C#N)C(=O)NC=C1 | |
| 150.14 | |
| 150.14 | |
| Crystalline Powder |
| 21642-98-8 | |
| White-Yellow | |
| MFCD00975442 | |
| 3439 | |
| MWGIDWPSRDMIQN-UHFFFAOYSA-N | |
| 4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile | |
| 2786702 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 21642-98-8 | |
| 150.14 | |
| MWGIDWPSRDMIQN-UHFFFAOYSA-N | |
| 2786702 | |
| COC1=C(C#N)C(=O)NC=C1 |
| C7H6N2O2 | |
| MFCD00975442 | |
| 4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile, 4-methoxy-2-oxo-1,2-dihydro-pyridine-3-carbonitrile, 3-cyano-2-hydroxy-4-methoxypyridine, 4-methoxy-2-oxo-1,2-dihydro-3-pyridinecarbonitrile, 2-hydroxy-4-methoxynicotinonitrile, 1,2-dihydro-4-methoxy-2-oxo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4-methoxy-2-oxo, 3-cyano-4-methoxy-2 1h-pyridinone, 2-hydroxy-4-methoxy-nicotinonitrile, 3-cyano-4-methoxy-2-1h-pyridinone | |
| 4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile |
Safety and Handling
TSCA : No