Learn More
3-Cyano-4-methoxy-2-pyridone, 95%, Thermo Scientific Chemicals
$587.85 - $587.85
Chemical Identifiers
| CAS | 21642-98-8 |
|---|---|
| Molecular Formula | C7H6N2O2 |
| Molecular Weight (g/mol) | 150.14 |
| MDL Number | MFCD00975442 |
| InChI Key | MWGIDWPSRDMIQN-UHFFFAOYSA-N |
| Synonym | 4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile, 4-methoxy-2-oxo-1,2-dihydro-pyridine-3-carbonitrile, 3-cyano-2-hydroxy-4-methoxypyridine, 4-methoxy-2-oxo-1,2-dihydro-3-pyridinecarbonitrile, 2-hydroxy-4-methoxynicotinonitrile, 1,2-dihydro-4-methoxy-2-oxo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4-methoxy-2-oxo, 3-cyano-4-methoxy-2 1h-pyridinone, 2-hydroxy-4-methoxy-nicotinonitrile, 3-cyano-4-methoxy-2-1h-pyridinone |
| PubChem CID | 2786702 |
| SMILES | COC1=C(C#N)C(=O)NC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6427914
|
Thermo Scientific Chemicals
H6427914 |
25 g |
Each for $587.85
|
|
|||||
Description
Used as a intermediate for gimeracil and pharmaceutical application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a intermediate for gimeracil and pharmaceutical application.
Solubility
Soluble in Dimethyl sulfoxide.
Notes
Hydroscopic. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from water/moisture and oxidizing agent.
Chemical Identifiers
| 21642-98-8 | |
| 150.14 | |
| MWGIDWPSRDMIQN-UHFFFAOYSA-N | |
| 2786702 |
| C7H6N2O2 | |
| MFCD00975442 | |
| 4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile, 4-methoxy-2-oxo-1,2-dihydro-pyridine-3-carbonitrile, 3-cyano-2-hydroxy-4-methoxypyridine, 4-methoxy-2-oxo-1,2-dihydro-3-pyridinecarbonitrile, 2-hydroxy-4-methoxynicotinonitrile, 1,2-dihydro-4-methoxy-2-oxo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4-methoxy-2-oxo, 3-cyano-4-methoxy-2 1h-pyridinone, 2-hydroxy-4-methoxy-nicotinonitrile, 3-cyano-4-methoxy-2-1h-pyridinone | |
| COC1=C(C#N)C(=O)NC=C1 |
Specifications
| 21642-98-8 | |
| Yellow | |
| MFCD00975442 | |
| UN3439 | |
| Hygroscopic | |
| Soluble in Dimethyl sulfoxide. | |
| COC1=C(C#N)C(=O)NC=C1 | |
| 150.14 | |
| 150.14 | |
| Crystalline Solid |
| 278°C | |
| C7H6N2O2 | |
| 25 g | |
| 5929290 | |
| 4-methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile, 4-methoxy-2-oxo-1,2-dihydro-pyridine-3-carbonitrile, 3-cyano-2-hydroxy-4-methoxypyridine, 4-methoxy-2-oxo-1,2-dihydro-3-pyridinecarbonitrile, 2-hydroxy-4-methoxynicotinonitrile, 1,2-dihydro-4-methoxy-2-oxo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4-methoxy-2-oxo, 3-cyano-4-methoxy-2 1h-pyridinone, 2-hydroxy-4-methoxy-nicotinonitrile, 3-cyano-4-methoxy-2-1h-pyridinone | |
| MWGIDWPSRDMIQN-UHFFFAOYSA-N | |
| 4-methoxy-2-oxo-1H-pyridine-3-carbonitrile | |
| 2786702 | |
| 95% | |
| 3-Cyano-4-methoxy-2-pyridone |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only