missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-iodopyridine, 97%

Catalog No. AC437450010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC437450010 1 g
AC437450050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437450010 Supplier Thermo Scientific Chemicals Supplier No. 437450010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 490039-72-0 | C6H3IN2 | 230.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 490039-72-0
Molecular Formula C6H3IN2
Molecular Weight (g/mol) 230.01
InChI Key CJKHIIQONPNAIP-UHFFFAOYSA-N
Synonym 3-cyano-4-iodopyridine, 4-iodonicotinonitrile, 3-pyridinecarbonitrile,4-iodo, acmc-20aibr, 4-iodo-nicotinonitrile, 4-iodo-3-pyridinecarbonitrile, 4-iodanylpyridine-3-carbonitrile
PubChem CID 11138941
IUPAC Name 4-iodopyridine-3-carbonitrile
SMILES C1=CN=CC(=C1I)C#N

Specifications

Melting Point 114°C to 115°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 230.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyano-4-iodopyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh

RUO – Research Use Only