missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-hydroxybenzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog No. AAH66434MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66434MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66434MD Supplier Thermo Scientific Chemicals Supplier No. H66434MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 775351-56-9
Molecular Formula C13H16BNO3
Molecular Weight (g/mol) 245.085
MDL Number MFCD16994355
InChI Key PUOBFTBTFCPICR-UHFFFAOYSA-N
Synonym 2-hydroxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, benzonitrile, 2-hydroxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, acmc-209pam, 3-cyano-4-bydroxy-boronic acid pinacol ester, 2-hydroxy-benzonitrile-5-boronic acid pinacol ester, 3-cyano-4-hydroxyphenyl boronic acid pinacol ester, 2-hydroxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 2-hydroxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile
PubChem CID 11481958
IUPAC Name 2-hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)O)C#N

Specifications

Quantity 250 mg
Beilstein 10347892
Formula Weight 245.09
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Cyano-4-hydroxybenzeneboronic acid pinacol ester
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only