missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 96%

Catalog No. AAH6224506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6224506 5 g
AAH6224503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6224506 Supplier Thermo Scientific Chemicals Supplier No. H6224506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 775351-57-0 | C13H15BFNO2 | 247.076 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 775351-57-0
Molecular Formula C13H15BFNO2
Molecular Weight (g/mol) 247.076
MDL Number MFCD06795681
InChI Key RYJOVQGRIOURGT-UHFFFAOYSA-N
Synonym 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-4-fluorophenylboronic acid, pinacol ester, 3-cyano-4-fluorophenylboronic acid pinacol ester, benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-cyano-4-fluorobenzeneboronic acid pinacol ester, 3-cyano-4-fluorophenyl-boronic acid pinacol ester, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 2-fluoro-5-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzonitrile, 2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile
PubChem CID 11402361
IUPAC Name 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C#N

Specifications

Melting Point 75°C to 76°C
Quantity 5 g
Formula Weight 247.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Cyano-4-fluorobenzeneboronic acid pinacol ester
Hazard Category H301+H311+H331-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only