missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2-hydroxy-4,6-dimethylpyridine, 98%

Catalog No. AAA1246622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1246622 100 g
AAA1246614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1246622 Supplier Thermo Scientific Chemicals Supplier No. A1246622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 769-28-8 | C8H8N2O | 148.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 769-28-8
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
MDL Number MFCD00006269
InChI Key OCYMJCILWYHKAU-UHFFFAOYSA-N
Synonym 2-hydroxy-4,6-dimethylnicotinonitrile, 3-cyano-4,6-dimethyl-2-hydroxypyridine, 4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 2-hydroxy-4,6-dimethylpyridine-3-carbonitrile, unii-ots4s2r79e, 4,6-dimethyl-2-hydroxynicotinonitrile, 3-pyridinecarbonitrile, 1,2-dihydro-4,6-dimethyl-2-oxo, 3-cyano-2-hydroxy-4,6-dimethylpyridine, nicotinonitrile, 1,2-dihydro-4,6-dimethyl-2-oxo, ots4s2r79e
PubChem CID 69856
IUPAC Name 4,6-dimethyl-2-oxo-1H-pyridine-3-carbonitrile
SMILES CC1=CC(=C(C(=O)N1)C#N)C

Specifications

Melting Point ∼290°C (decomposition)
Quantity 100 g
Beilstein 130992
Formula Weight 148.17
Percent Purity 98%
Chemical Name or Material 3-Cyano-2-hydroxy-4,6-dimethylpyridine
Hazard Category H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
EINECSNumber 212-207-5
RTECSNumber QT3046500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only