missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyano-2-fluoropyridine 98.0+%, TCI America™
$93.21 - $348.57
Chemical Identifiers
| CAS | 3939-13-7 |
|---|---|
| Molecular Formula | C6H3FN2 |
| Molecular Weight (g/mol) | 122.102 |
| MDL Number | MFCD03095082 |
| InChI Key | USIDQCCXMGJOJM-UHFFFAOYSA-N |
| Synonym | 3-cyano-2-fluoropyridine, 2-fluoronicotinonitrile, 2-fluoro-3-cyanopyridine, 3-pyridinecarbonitrile, 2-fluoro, 2-fluoro-nicotinonitrile, 2-fluoro-3-pyridinecarbonitrile, pubchem1108, 2-fluoronicotin-onitrile, acmc-1ckgi, 2-fluoro-3-nitrylpyridine |
| PubChem CID | 2783254 |
| IUPAC Name | 2-fluoropyridine-3-carbonitrile |
| SMILES | C1=CC(=C(N=C1)F)C#N |
Chemical Identifiers
| 3939-13-7 | |
| 122.102 | |
| USIDQCCXMGJOJM-UHFFFAOYSA-N | |
| 2783254 | |
| C1=CC(=C(N=C1)F)C#N |
| C6H3FN2 | |
| MFCD03095082 | |
| 3-cyano-2-fluoropyridine, 2-fluoronicotinonitrile, 2-fluoro-3-cyanopyridine, 3-pyridinecarbonitrile, 2-fluoro, 2-fluoro-nicotinonitrile, 2-fluoro-3-pyridinecarbonitrile, pubchem1108, 2-fluoronicotin-onitrile, acmc-1ckgi, 2-fluoro-3-nitrylpyridine | |
| 2-fluoropyridine-3-carbonitrile |
Specifications
| 3939-13-7 | |
| C6H3FN2 | |
| 5 g | |
| USIDQCCXMGJOJM-UHFFFAOYSA-N | |
| 2-fluoropyridine-3-carbonitrile | |
| 2783254 | |
| ≥98.0% (GC) | |
| 3-Cyano-2-fluoropyridine |
| Green-White | |
| MFCD03095082 | |
| 3-cyano-2-fluoropyridine, 2-fluoronicotinonitrile, 2-fluoro-3-cyanopyridine, 3-pyridinecarbonitrile, 2-fluoro, 2-fluoro-nicotinonitrile, 2-fluoro-3-pyridinecarbonitrile, pubchem1108, 2-fluoronicotin-onitrile, acmc-1ckgi, 2-fluoro-3-nitrylpyridine | |
| C1=CC(=C(N=C1)F)C#N | |
| 122.102 | |
| 122.10 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No