missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2-fluoropyridine, 98%

Catalog No. AAL1953803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1953803 1 g
AAL1953806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1953803 Supplier Thermo Scientific Chemicals Supplier No. L1953803

CAS: 3939-13-7 | C6H3FN2 | 122.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3939-13-7
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD03095082
InChI Key USIDQCCXMGJOJM-UHFFFAOYSA-N
Synonym 3-cyano-2-fluoropyridine, 2-fluoronicotinonitrile, 2-fluoro-3-cyanopyridine, 3-pyridinecarbonitrile, 2-fluoro, 2-fluoro-nicotinonitrile, 2-fluoro-3-pyridinecarbonitrile, pubchem1108, 2-fluoronicotin-onitrile, acmc-1ckgi, 2-fluoro-3-nitrylpyridine
PubChem CID 2783254
IUPAC Name 2-fluoropyridine-3-carbonitrile
SMILES C1=CC(=C(N=C1)F)C#N

Specifications

Melting Point 27°C to 30°C
Quantity 1 g
Beilstein 386567
Formula Weight 122.1
Percent Purity 98%
Chemical Name or Material 3-Cyano-2-fluoropyridine
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only