missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C32141G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C32141G Supplier TCI America Supplier No. C32141G
Only null left

Chemical Identifiers

CAS 957121-05-0
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD07374984
InChI Key HENIWPFEWBREIB-UHFFFAOYSA-N
Synonym 3-Cyano-2-fluorobenzeneboronic Acid
PubChem CID 44558173
IUPAC Name (3-cyano-2-fluorophenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)C#N)F)(O)O

Specifications

Color White
Quantity 1 g
Formula Weight 164.93
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyano-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)